Learn More
Invitrogen™ EDAC, 1-Ethyl-3-(3-Dimethylaminopropyl)carbodiimide, Hydrochloride
EDAC is a water-soluble carbodiimide that can be used to crosslink biological substances that contain carboxylate acids and primary amines.
Brand: Invitrogen™ E2247
Code : 50
Additional Details : Weight : 0.00454kg
Description
Protocols for crosslinking molecules with this reagent can be found in Bioconjugate Techniques, by G.T. Hermanson (B-7884).Specifications
191.7 | |
Solid |
For Research Use Only. Not for use in diagnostic procedures.