missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDG4 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577477
Additional Details : Weight : 0.01000kg
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Edg4 is a Lysophospholipid/Lysosphingolipid Receptor activated by phosphatidic acid (PA) and lysophosphatidic acid (LPA). It enhances the transcription of serum response element (SRE) reporter genes. In CD4(+) T cells, Edg4 may be involved in the control of interleukin-2 secretion. Edg4 expression has been documented in leukocytes, pancreas, spleen, thymus, prostate, and testis. Edg4 expression has also been seen in many cancer cell lines, including those of brain, blood, breast, ovary, cervix, colon, skin, and thyroid. ESTs have been isolated from B-cell/lung/testis, blood, brain, embryo, heart/melanocyte/uterus, liver, placenta, prostate, and skin libraries.Specifications
EDG4 (extracellular) | |
Polyclonal | |
Unconjugated | |
LPAR2 | |
LPA2 Receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
498609, 53978, 9170 | |
-20°C | |
Lyophilized |
Western Blot | |
0.85 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
Q9HBW0, Q9JL06 | |
LPAR2 | |
Peptide (C)NSGKELSLHWRPKD, corresponding to amino acid residues 16-29 of mouse LPA2 Receptor | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |