missing translation for 'onlineSavingsMsg'
Learn More

Ansell EDGE™ 40-400 Porous Nitrile Coated Medium Duty Cotton Gloves

Product Code. 12660892
Click to view available options
Size:
8
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Porous nitrile coating helps channel oil away from the glove's surface for a firmer grip in slightly oily applications. Glove is designed to mold to the hand for improved movement and dexterity.

  • Ergonomic, light, flexible, and tough
  • Porous nitrile coating helps channel oil away from the glove's surface for a firm grip
  • Designed to mold to the hand for improved movement and dexterity
  • Ideal for quick and accurate handling
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Coated Area Palm, Fingers
Construction Cut and Sewn
Coating Material Nitrile Foam
Color Natural
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Material Cotton
Product Type Dip-coated Gloves
Size 10
For Use With (Application) Automotive, Machinery/Equipment, and Metal Fabrication
Glove Cuff Style Knit
Series 40-400
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'