missing translation for 'onlineSavingsMsg'
Learn More

EDTA (Disodium Salt) 0.1 M (0.2 N) Analytical Volumetric Solution (AVL), Reagecon
BIOPHARMA

Fully factorized, high purity, stable, certified product, developed and tested for complexometric titrations.

Brand:  Reagecon ED2011

Product Code. 11576753

  • £35.90 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
  • Accurate, stable, reproducible and fully factorized.
  • Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
  • Stability is proven (either opened or unopened) for the shelf life of this product.
  • Certified by Reagecon’s ISO 17025 Accredited Test Method (INAB Ref: 264T)
  • Certified to a specification of ± 0.1%.
  • Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
  • May be used for manual titrations, or automatic titrations, to determine the end points by detection of any of several properties.
  • Complexometric titrimetry is based on the reaction of a metal ion with a ligand to form a complex.
  • Methods have been developed, validated and published for detection or quantification of almost every metal in the periodic table using EDTA complexation.
Specifications

Specifications

Accuracy ± 0.1%
HDPE Bottle GL45
Titration
0.1 M (0.2 N)
1 L
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers