missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA, Disodium Salt, Dihydrate, Crystal, Ultrapure Bioreagent, J.T. Baker™
High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography
£164.00 - £1365.00
Chemical Identifiers
CAS | 6381-92-6 |
---|---|
Molecular Formula | C10H18N2Na2O10 |
Molecular Weight (g/mol) | 372.24 |
MDL Number | MFCD00150037,MFCD00003541 |
InChI Key | OVBJJZOQPCKUOR-UHFFFAOYSA-L |
Synonym | edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs |
PubChem CID | 44120005 |
SMILES | O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15598214
|
Avantor J.T.Baker
4040-00 |
100 g | Poly Bottle |
£164.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15549660
|
Avantor J.T.Baker
4040-01 |
500 g | Poly Bottle |
£355.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15508224
|
Avantor J.T.Baker
4040-04 |
1 kg | Poly Bottle |
£397.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15518224
|
Avantor J.T.Baker
4040-06 |
5 kg | Poly Pail |
£1365.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For Molecular Biology Applications
Chemical Identifiers
6381-92-6 | |
372.24 | |
OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
44120005 |
C10H18N2Na2O10 | |
MFCD00150037,MFCD00003541 | |
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
Specifications
6381-92-6 | |
MFCD00150037,MFCD00003541 | |
OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate | |
44120005 | |
Ultrapure Bioreagent | |
EDTA, Disodium Salt, Dihydrate |
C10H18N2Na2O10 | |
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
372.24 | |
372.24 | |
Crystals |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title