missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid tetrasodium salt dihydrate, (EDTA), 99-101%, Ultrapure

Product Code. 15805288 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 kg
25 kg
Packaging:
Plastic bottle
Unit Size:
1kg
25kg
This item is not returnable. View return policy

Product Code. 15805288

Brand: Thermo Scientific J15700.C4

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10378-23-1 | C10H20N2Na4O10 | 420.233 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10378-23-1
Molecular Formula C10H20N2Na4O10
Molecular Weight (g/mol) 420.233
MDL Number MFCD00150026
InChI Key DGWOWDJDIBPYOR-UHFFFAOYSA-N
Synonym ethylenediaminetetraacetic acid tetrasodium salt dihydrate, tetrasodium ethylenediaminetetraacetate dihydrate, edta-na4, edta tetrasodium salt, edta tetrasodium salt hydrate, ethylenediaminetetraacetic acid tetrasodium salt hydrate, 64-02-8 anhydrous, ksc909a7h, edta tetrasodium salt dihydrate, tetrapotassium ion 4-edta dihydrate
PubChem CID 131843362
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;sodium;dihydrate
SMILES C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O.O.[Na].[Na].[Na].[Na]

Specifications

Physical Form Crystalline Powder
Quantity 25 kg
Beilstein 3861753
Formula Weight 416.2
Percent Purity 99 to 101%
Packaging Plastic bottle
Chemical Name or Material Ethylenediaminetetraacetic acid tetrasodium salt dihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.