Learn More
Edwards Fittings for BOC High Vacuum Pumps: Blank Flange
For use with BOC high vacuum pumps
Brand:  Edwards C10514368
 View more versions of this product
Product Code. 12925264
  • £13.00 / Each
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made of aluminum
  • Aluminum is ideal for achieving dependable cost-effective performance down to 10-7mbar
Specifications
Specifications
BOC high vacuum pump
Aluminum
40 x 26.2 x 6 x 2.5 mm
NW25 Blank Flange
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Edwards Fittings for BOC High Vacuum Pumps: Blank Flange > NW25 Blank flange