Learn More
Cole-Parmer™ Edwards™ Vacuum Pump Fitting
Adapters fit almost every kind of vacuum pump and accessory
Brand: Cole-Parmer™ 31401-39
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Vacuum pump fittings.
- Stainless steel construction.
- For use with a wide range of applications.
Specifications
Vacuum Pump Fitting, 316L SS adapter, NW 25 to 1/4in NPT(F) | |
Vacuum Flange x NPT(F) | |
Stainless Steel | |
1/4 in. |