missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ EED226, 95%
Brand: Thermo Scientific™ 468141000
370.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
EED226 | |
N-(2-Furanylmethyl)-8-[4-(methylsulfonyl)phenyl]-1,2,4-triazolo[4,3-c]pyrimidin-5-amine | |
C17 H15 N5 O3 S | |
Solid | |
95% |
2083627-02-3 | |
100 mg | |
369.40 | |
Reagent |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
344414 | Certificate of Analysis | 09/10/2020 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ EED226, 95% > 100 mg
Spot an opportunity for improvement?Share a Content Correction