Learn More
Thermo Scientific Chemicals Efaproxiral sodium, 98%
£80.10 - £293.00
Chemical Identifiers
CAS | 170787-99-2 |
---|---|
Molecular Formula | C20H22NNaO4 |
Molecular Weight (g/mol) | 363.38 |
InChI Key | SWDPIHPGORBMFR-UHFFFAOYSA-M |
Synonym | efaproxiral sodium, sodium 2-4-2-3,5-dimethylphenyl amino-2-oxoethyl phenoxy-2-methylpropanoate, rsr 13 sodium, unii-3l83qp52xi, rsr13 sodium, efaproxiral sodium usan, rsr-13 |
PubChem CID | 2725048 |
IUPAC Name | sodium;2-[4-[2-(3,5-dimethylanilino)-2-oxoethyl]phenoxy]-2-methylpropanoate |
SMILES | CC1=CC(=CC(=C1)NC(=O)CC2=CC=C(C=C2)OC(C)(C)C(=O)[O-])C.[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15556006
|
Thermo Scientific Chemicals
460102500 |
250 mg |
£80.10
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15546006
|
Thermo Scientific Chemicals
460100010 |
1 g |
£293.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
170787-99-2 | |
363.38 | |
efaproxiral sodium, sodium 2-4-2-3,5-dimethylphenyl amino-2-oxoethyl phenoxy-2-methylpropanoate, rsr 13 sodium, unii-3l83qp52xi, rsr13 sodium, efaproxiral sodium usan, rsr-13 | |
sodium;2-[4-[2-(3,5-dimethylanilino)-2-oxoethyl]phenoxy]-2-methylpropanoate |
C20H22NNaO4 | |
SWDPIHPGORBMFR-UHFFFAOYSA-M | |
2725048 | |
CC1=CC(=CC(=C1)NC(=O)CC2=CC=C(C=C2)OC(C)(C)C(=O)[O-])C.[Na+] |
Specifications
170787-99-2 | |
Authentic | |
C20H22NNaO4 | |
SWDPIHPGORBMFR-UHFFFAOYSA-M | |
sodium;2-[4-[2-(3,5-dimethylanilino)-2-oxoethyl]phenoxy]-2-methylpropanoate | |
2725048 | |
98% |
13% max. | |
97.5% min. (HPLC) | |
efaproxiral sodium, sodium 2-4-2-3,5-dimethylphenyl amino-2-oxoethyl phenoxy-2-methylpropanoate, rsr 13 sodium, unii-3l83qp52xi, rsr13 sodium, efaproxiral sodium usan, rsr-13 | |
CC1=CC(=CC(=C1)NC(=O)CC2=CC=C(C=C2)OC(C)(C)C(=O)[O-])C.[Na+] | |
363.38 | |
363.38 | |
Efaproxiral sodium |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
- Efaproxiral sodium
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only