Learn More
Manutan Egg link for 10mm chain
Brand: Manutan A341457
Code : Z3
Additional Details : Weight : 1.00000kg
Description
A range of high strength chain and fittings to suit a variety of lifting, lashing, suspending or security applications. Fittings - 'connector' for linking components and/or chain consists of 2 matched halves joined by a cross pin and tension bush. Other fittings are connected to chain using the Kuplex clevis system with load retaining pin. Test certificate supplied- Egg link
- Chain diameter: 10mm
- Workload Limit: 3200