missing translation for 'onlineSavingsMsg'
Learn More

Melford EGTA 0.5M Solution pH 8.0

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

Brand:  Melford E14100-50.0

Product Code. 30018249

This item is not returnable. View return policy

Description

Description

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

  • Soluble in: Water
  • Store at: 2-8°C
Specifications

Specifications

EGTA 0.5 M Solution pH 8.0
67-42-5, 7732-18-5
50 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers