missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGTA, Tocris Bioscience™
Click to view available options
Quantity:
1 g
Description
Calcium chelator; protects against cell death caused by nitric oxide-induced calcium influx into nerve cells.
Specifications
CAS | 67-42-5 |
Molecular Formula | C14H24N2O10 |
Quantity | 1 g |
Synonym | egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn |
InChI Key | DEFVIWRASFVYLL-UHFFFAOYSA-N |
SMILES | C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O |
IUPAC Name | 2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid |
Molecular Weight (g/mol) | 380.35 |
PubChem CID | 6207 |
ChEBI | CHEBI:30740 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'