missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EI24 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | EI24 |
---|---|
Concentration | 0.17 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16810005
|
Proteintech
20456-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16800005
|
Proteintech
20456-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a serine/threonine-protein kinase that functions in cell division. Together with the kinesin KIF14, this protein localizes to the central spindle and midbody, and functions to promote efficient cytokinesis. This protein is involved in central nervous system development. Polymorphisms in this gene are associated with bipolar disorder and risk for schizophrenia. Alternative splicing results in multiple transcript variants.Specifications
EI24 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
O14681, Q4KM77, Q61070 | |
13663, 300514, 9538 | |
EI24 Fusion Protein Ag14169 | |
Primary | |
-20°C | |
EI24 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
EI24, etoposide induced 2.4 mRNA, p53 induced gene 8 protein, p53-induced factor Ei24, PIG8, TP53I8 | |
EI24 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |