Learn More
Eicosane, 99%, Thermo Scientific Chemicals
£14.57 - £182.91
Chemical Identifiers
CAS | 112-95-8 |
---|---|
Molecular Formula | C20H42 |
Molecular Weight (g/mol) | 282.56 |
MDL Number | MFCD00009344 |
InChI Key | CBFCDTFDPHXCNY-UHFFFAOYSA-N |
Synonym | eicosane, n-eicosane, unii-3aya9kec48, ccris 663, nonadecane, methyl, 3aya9kec48, eicosane, analytical standard, octyldodecane, n-icosane |
PubChem CID | 8222 |
ChEBI | CHEBI:43619 |
IUPAC Name | icosane |
SMILES | CCCCCCCCCCCCCCCCCCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10246640
|
Thermo Scientific Acros
204370020 |
2 g | Glass bottle |
£14.57
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10144410
|
Thermo Scientific Acros
204371000 |
100 g | Glass bottle |
£43.72
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10636734
|
Thermo Scientific Acros
204375000 |
500 g | Glass bottle |
£182.91
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-95-8 | |
282.56 | |
CBFCDTFDPHXCNY-UHFFFAOYSA-N | |
8222 | |
icosane |
C20H42 | |
MFCD00009344 | |
eicosane, n-eicosane, unii-3aya9kec48, ccris 663, nonadecane, methyl, 3aya9kec48, eicosane, analytical standard, octyldodecane, n-icosane | |
CHEBI:43619 | |
CCCCCCCCCCCCCCCCCCCC |
Specifications
112-95-8 | |
White | |
220.0°C (30.0 mmHg) | |
Authentic | |
C20H42 | |
MFCD00009344 | |
0.7886 | |
Solubility in water: insoluble. Other solubilities: soluble in ether | |
CCCCCCCCCCCCCCCCCCCC | |
282.56 | |
CHEBI:43619 | |
99% | |
Eicosane |
36.0°C to 38.0°C | |
0.7886g/mL | |
>112°C | |
98.5% min. (GC) | |
CH3(CH2)18CH3 | |
01, 174 | |
eicosane, n-eicosane, unii-3aya9kec48, ccris 663, nonadecane, methyl, 3aya9kec48, eicosane, analytical standard, octyldodecane, n-icosane | |
CBFCDTFDPHXCNY-UHFFFAOYSA-N | |
icosane | |
8222 | |
282.54 | |
Crystalline Solid |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 204-018-1
- Eicosane
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.