Learn More
eIF3b Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541773
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: YRKMAQELYM EQKNERLELR GGVDTDELDS NVDDWEEETI EFFVTEEIIP Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
eIF3b Polyclonal antibody specifically detects eIF3b in Human samples. It is validated for Western Blot.Specifications
eIF3b | |
Polyclonal | |
Unconjugated | |
EIF3B | |
AL033316; AL033334; AL033369; AW208965; D5Wsu45e; eIF3 p110; eIF3 p116; EIF3B; eIF3b {ECO:0000255; eIF-3-eta; EIF3-ETA; eIF-3-eta {ECO:0000255; EIF3-P110; EIF3-P116; EIF3S9; eukaryotic translation initiation factor 3 subunit 9; eukaryotic translation initiation factor 3 subunit 9 {ECO:0000255; Eukaryotic translation initiation factor 3 subunit B; eukaryotic translation initiation factor 3 subunit B {ECO:0000255; eukaryotic translation initiation factor 3, subunit 9 (eta); eukaryotic translation initiation factor 3, subunit 9 (eta, 116kD); eukaryotic translation initiation factor 3, subunit 9 eta, 116kDa; eukaryotic translation initiation factor 3, subunit B; HAMAP-Rule:MF_03001}; hPrt1; MGC104664; MGC131875; PRT1; Prt1 homolog | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8662 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P55884 | |
EIF3B | |
Synthetic peptide directed towards the C-terminal of human EIF3S9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |