missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4A2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523264
Code : LD
Additional Details : Weight : 0.01000kg
Description
eIF4A2 is a 407 amino acid ATP-dependent RNA helicase belonging to the DEAD box helicase family. It contains one helicase ATP-binding domain and a helicase C-terminal domain. eIF4A2, a subunit of the eIF4F complex, acts as a protein-synthesis initiation factor and thus plays an important role in the binding of mRNA to the 43S pre-initiation complex of ribosomes. Human eIF4A2 interacts with the NS5B protein of Hepatitis C virus (HCV) and facilitates the genomic RNA synthesis of NS5B protein by unwinding the secondary structure of the HCV genome. It is ubiquitously expressed in most of normal tissues, but in variable amounts.Specifications
eIF4A2 | |
Polyclonal | |
Unconjugated | |
EIF4A2 | |
4833432N07Rik; ATP dependent RNA helicase eIF4A 2; ATP-dependent RNA helicase eIF4A-2; BM-010; DDX2B; Eif4; EIF4A; EIF4A1; EIF4A2; EIF4AII; eIF-4A-II; eIF4A-II; EIF4F; Eukaryotic initiation factor 4A-II; eukaryotic translation initiation factor 4A; eukaryotic translation initiation factor 4A isoform 2; eukaryotic translation initiation factor 4A, isoform 2; eukaryotic translation initiation factor 4A1; eukaryotic translation initiation factor 4A2; hypothetical protein; N-terminally processed; protein synthesis initiation factor 4AII; RCJMB04_14a6; translational eukaryotic inititation factor 4AII | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13682, 1974, 395232, 488118 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
P10630, Q14240, Q8JFP1 | |
EIF4A2 | |
A portion of amino acids 50-100 of human eIF4A2 | |
100 μg | |
Primary | |
Canine, Chicken, Human, Mouse, Non-human Primate | |
Antibody | |
IgG |