Learn More
eIF4E Monoclonal Antibody (5D11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA1089
Code : PN
Additional Details : Weight : 0.01000kg
Description
MA1-089 detects eIF4E in human and mouse samples and has been successfully used in Western blot, immunofluorescence, immunocytochemistry and immunoprecipitation applications. MA1-089 does not react with rat eIF4E. The MA1-089 immunogen is full length human eIF4E produced in HEK293T cells.
eIF4E is involved in several cellular processes including enhanced translational efficiency, splicing, mRNA stability, and RNA nuclear export. All eukaryotic cellular mRNAs are blocked at their 5-prime ends with the 7-methylguanosine cap structure, m7GpppX (where X is any nucleotide). EIF4E is a eukaryotic translation initiation factor involved in directing ribosomes to the cap structure of mRNAs. It is a 24-kD polypeptide that exists as both a free form and as part of a multiprotein complex termed EIF4F. The EIF4E polypeptide is the rate-limiting component of the eukaryotic translation apparatus and is involved in the mRNA-ribosome binding step of eukaryotic protein synthesis. The other subunits of EIF4F are a 50-kD polypeptide, termed EIF4A, that possesses ATPase and RNA helicase activities, and a 220-kD polypeptide, EIF4G.Specifications
eIF4E | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
P06730, P63073 | |
Eif4e | |
Full length human eIF4E produced in HEK293T cells | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunoprecipitation, Immunoprecipitation, Western Blot | |
5D11 | |
Unconjugated | |
Eif4e | |
AUTS19; CBP; EG668879; EIF4E; EIF-4E; EIF4E1; EIF4EL1; Eif4e-ps; EIF4F; eIF-4F 25 kDa subunit; eukaryotic translation initiation factor 4E; eukaryotic translation initiation factor 4E-like 1; eukaryotic translation initiation factor 4E-like protein; eukaryotic translation initiation factor 4-like protein; If4e; MGC111573; mRNA cap-binding protein; OTTHUMP00000219703; OTTHUMP00000219704; OTTHUMP00000219705; translation initiation factor eIF-4E | |
Mouse | |
Protein A | |
RUO | |
13684, 1977 | |
-20°C | |
Liquid |