missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF4E2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12227-1-AP-20UL
This item is not returnable.
View return policy
Description
Recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structures.Specifications
EIF4E2 | |
Polyclonal | |
Unconjugated | |
EIF4E2 | |
4E LP, 4EHP, eIF 4E type 2, eIF4E like protein 4E LP, eIF4E type 2, EIF4E2, EIF4EL3, IF4e, mRNA cap binding protein 4EHP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26987, 363275, 9470 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60573, Q8BMB3 | |
Eif4e2 | |
EIF4E2 Fusion Protein Ag2864 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction