Learn More
eIF4ENIF1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518235
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
The protein encoded by this gene is a nucleocytoplasmic shuttle protein for the translation initiation factor eIF4E. This shuttle protein interacts with the importin alpha-beta complex to mediate nuclear import of eIF4E. It is predominantly cytoplasmic; its own nuclear import is regulated by a nuclear localization signal and nuclear export signals. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
eIF4ENIF1 | |
Polyclonal | |
Unconjugated | |
EIF4ENIF1 | |
2610509L04Rik; 4E-T; A930019J01Rik; AA410001; AU021239; Clast4; D11Ertd166e; eIF4E transporter; EIF4ENIF1; eIF4E-transporter; Eukaryotic translation initiation factor 4E nuclear import factor 1; eukaryotic translation initiation factor 4E transporter; Eukaryotic translation initiation factor 4E transporter-like protein; FLJ21601; FLJ26551; OTTHUMP00000063276; RGD1560908 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
56478 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9NRA8 | |
EIF4ENIF1 | |
Synthetic peptide sequence (AKVISVDELEYRQ) corresponding to the C-terminus amino acids of EIF4ENIF1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |