Learn More
Eighteen element solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand:  Fisher Chemical
 View more versions of this product
Product Code. 12936193
  • £375.00 / 50mL
Explore more special offers
Description
Description
Eighteen elements solution - Ag, Al, As, Ba, Be, Cd, Cr, Co, Cu, Mn, Ni, Pb Se, Th, Tl, U, V, Zn for ICP/MS 10mg/L HNO3 matrix
Specifications
Specifications
Eighteen Elements
Bottle
ICP/MS Calibration Multi-element Standard Solution
2% HNO3
10mg/L
50 mL
Ag,Al,As,Ba,Be,Cd,Cr,Co,Cu,Mn,Ni,Pb,Se,Th,Tl,U,V,Zn
Liquid
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Multi-element Standard Solution for ICP. 18 Components in HNO3 2% (MS 008)
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to .