missing translation for 'onlineSavingsMsg'
Learn More

A&D Weighing EK-AEP Intrinsically Safe Portable Balances ATEX

A&D Weighing EK-AEP Intrinsically Safe Portable Balances ATEX feature a compact rugged design, reliable SG weighing technology, and versatile weighing ranges.

Brand:  A&D Weighing EK-12KAEP

1334.14 GBP valid until 2024-12-31
Use promo code "21987" to get your promotional price.



Additional Details : Weight : 9.00000kg

 View more versions of this product

Product Code. 16874591

  • £1426.12 / Each
Estimated Shipment: 14-08-2024
to see stock.

Description

Description

  • Compact, lightweight and extremely affordable while providing excellent explosion protection
  • Equipped with an optical connector for data output via optical fiber cable and an AD-1611 optical communication adapter (both sold separately)
  • Battery operation (o be used only with four DURACELL AA alkaline MN1500 LR6, ENERGIZER AA E91 alkaline LR6 AM3, or Panasonic AA alkaline LR6 (XJ) 1.5 V batteries) (approx. 250 hours) for optimal portability without the need for complicated wiring via a safety barrier
  • GMP/GLP compliant output to meet documentation requirements
  • Easy-to-read LCD display with backlight (16 mm character height)
  • Auto power OFF function to save battery power
  • Hygienic and chemically resistant stainless steel (SUS304) weighing pan
  • Multiple weighing units: g, oz, lb, ozt, ct, mom, dwt, gr, N, in addition to SG (specific gravity)
  • Counting mode with Automatic Counting Accuracy Improvement (ACAI) functionality
  • Percent mode function for target weighing, checking the sample variation, etc.
  • Comparator function for faster and more accurate filling, packing, sorting, etc.
  • Response characteristics adjustable depending on the degree of drafts and vibrations at the locationSecurity ring for wire-locking the balance to prevent theft
Specifications

Specifications

GMP/GLP/ISO/IECEx/ATEX/NEC505: Class I, Zone 0, AEx/Ex ia op is IIB T3, NEC500: IS/I/1/C,D/T3C
12 kg
1 g
EK-12KAEP
Fibre Optic Interface (requires AD-1611 and Fibre Optic Cable)
Backlit LCD
ABS, Stainless Steel
To be used only with four DURACELL AA alkaline MN1500 LR6, ENERGIZER AA E91 alkaline LR6 AM3, or Panasonic AA alkaline LR6 (XJ) 1.5 V batteries
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers