Learn More
Chemsplash Eka Trouser Product Code.: 30058436
Estimated Shipment: 28-11-2025
to see stock.
Add to basket
Chemsplash Eka Trouser
GREENER_CHOICE
Product Code. 30058436
2X-Large
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Product Code. 30058436
Brand: Chemsplash 2616.400.008
Estimated Shipment: 28-11-2025
to see stock.
Add to basket
Add to basket
Trousers with Elasticated Waist and Ankles are strong and resistant to chemical splash and dust.
Chemsplash Trousers with Elasticated Waist and Ankles
  • Chemsplash 55GSM Type PB 6B Fabric
  • Elasticated Waist and Ankles
  • Silicone and Latex Free
  • Anti-Static Material
  • Non-Linting Fabric
  • PFAS Free
  • Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9
Description: Chemsplash Trousers with Elasticated Waist and Ankles are made of soft and microporous Chemsplash fabric. They are strong and resistant to chemical splash and dust, lint-free, and feature anti-static properties under EN1149-5:2008. Chemsplash accessories are designed to protect the wearer from the environment or the environment from the wearer. This range has been crafted to offer maximum comfort and wearability. The trousers conform to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9 standards.
Greener Choice Claims
  • Environmental benefits include:
    • Recycled Content
Specifications
Size 2X-Large
Gender Unisex
Product Type Trouser
Material Multi-layer Barrier Laminate Fabric
Color White
Ankle Style Elastic
Seam Type Stitched
No. of Pockets None
Disposable Single-use
Latex Free Yes
Sterility Non-sterile
Product Title
Chemsplash Eka Trouser >