missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Elastic Sleeves for CoolRack Modules
BIOPHARMA

For use with CoolRack Modules. Corning™ Elastic Sleeves for CoolRack Modules provides comfortable grip when handling cold, warm or wet CoolRack tube modules.

Brand:  Corning™ 432092

Product Code. 15555535

  • £26.35 / Pack of 4
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Colorful elastic sleeves to color-code experiments
  • Compatible with ice, dry ice and water baths
  • Four colors: purple, green, orange, pink

 

Sleeves fit the following modules:

  • All 15-well CoolRack modules
  • All 30-well CoolRack modules
  • CoolRack 15mL
  • CoolRack 50mL
  • CoolRack VS13
  • CoolRack V13
  • CoolRack V16
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Green, Orange, Purple, Pink
CoolRack Modules
Elastic Sleeves
Color-coding CoolRack tube modules
4/pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.