missing translation for 'onlineSavingsMsg'
Learn More
Euro Protection™ Elastomer Caps Related Noise Earplug Product Code.: 12067816

Euro Protection™ Elastomer Caps Related Noise Earplug

Product Code. 12067816
Color: Yellow/Blue
Click to view available options
Color: Yellow/Blue
This item is not returnable. View return policy

Product Code. 12067816

Brand: Euro Protection™ 30213

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Order Info

dimProductType

Specifications

Color Yellow, Blue
Certifications/Compliance EC Certificate of Type1390 Issued by INSPEC
Cord Material Elastomer
For Use With (Application) Noisy Industrial Environments
Quantity 12Box
Product Type Caps Related Noise Earplug
Material Elastomer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Euro Protection™ Elastomer Caps Related Noise Earplug >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.