missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ Electrical Component IO90 K3

Mains connection 400 V for ION-CLASSIC-90 model IO90.195.120.K3.WDC

Brand:  Asecos™ 38038

Product Code. 16319426

  • £115.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

For indoor use only

This item is not returnable. View return policy

Description

Description

  • Electrical component
  • For model: IO90.195.120.K3.WDC
  • For tall cabinets with body width of 1200 cm
Specifications

Specifications

ION-CLASSIC-90 model IO90.195.120.K3.WDC
Electrical component mains connection 400 V
Electrical component
Product Suggestions

Product Suggestions

Videos

Videos

  • asecos ION-LINE

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Asecos™ Electrical Component IO90 K3 > Description: Electrical component mains connection 400 V

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.