Learn More
Kimberly-Clark Professional™ Electronic Cassette Dispenser Floor Stand
Kimberly-Clark™ Electronic Cassette Dispenser Floor Stand is compatible with a variety of electronic skin care dispensers, so you can offer hand sanitizer wherever you most need it.
Brand: Kimberly-Clark Professional™ 11430
175.28 GBP valid until 2024-12-31
Use promo code "22916" to get your promotional price.
Code : NEW
Additional Details : Weight : 6.63300kg
Description
- Portable, adjustable and durable, making it a smart option for break rooms, restrooms, entryways and much more
- Choose from a wide variety of skincare and hand sanitizer cassettes from the trusted Kleenex™ Brand
- Use with a variety of compatible Electronic Skin Care Dispensers, sold separately
Specifications
1.46 m |