Learn More
Fisherbrand™ Electrode Connection Adapter
Use with metallic electrodes having pin connectors. Fisherbrand™ Electrode Connection Adapter is a single pin plug that connects directly to a BNC jack.
Brand: Fisherbrand™
Code : 43
Additional Details : Weight : 0.10000kg
Description
Use with metallic electrodes having pin connectors. Fisherbrand™ Electrode Connection Adapter is a single pin plug that connects directly to a BNC jack.
- Pin plug to BNC input jack
- Use with metallic electrodes having pin connectors
Specifications
Pin Plug | |
BNC Input Jack | |
Electrode Connection Adapter |