missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Electrode Storage Solution

For use in storage of pH and ORP electrodes. Hanna Instruments™ Electrode Storage Solution is prepared with reagent grade chemicals that can be used to ensure optimum performance of pH and ORP electrodes.

Brand:  Hanna Instruments™ HI70300M

Product Code. 10758891

  • £103.00 / 230mL
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Specifically formulated to minimize microbial growth and to prevent any diffusion/osmotic effects
  • Keep junction clear
  • Ensure fast and reliable measurement
  • To ensure optimum response time, glass sensor tip and reference junction of pH electrode should be kept moist and not be allowed to dry out when not in use
  • Store the electrode by placing electrode in small glass filled with storage solution or replacing solution in protective cap
Specifications

Specifications

230 mL
Storage
Bottle
pH and ORP Electrodes
Clear
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers