missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Electrolyte Fluoro Borate ISE

Brand:  Mettler Toledo™ 51107890

Product Code. 11947721

  • £50.45 / 20mL
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Electrolyte for Fluoroborate ISE, 20mL
Electrolyte
20 mL
20 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers