missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Electrolyte KCl Saturated with AgCl, Mettler Toledo™
Click to view available options
Quantity:
250 mL
Specifications
Specifications
Color | Colorless |
Description | Electrol. 3 mol/L KCl Sat AgCl, 6x250mL |
pH | 6 |
Physical Form | Liquid |
Quantity | 250 mL |
Solution Type | Electrolyte |
Packaging | 6x 250 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction