missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Electrolyte Standard Solution, 3M, Chem-Lab

Product Code. 11910049
250 mL
Click to view available options
Quantity:
250 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Enhance the performance and extend the life of electrodes. Potassium Chloride Electrolyte Standard Solution, 3M, Chem-Lab is a filling solution for pH electrodes, containing 224g KCl per liter of water. It is used for proper electrode storage to maintain optimal electrode performance.

When not in use, the glass tip of an electrode should be kept wet to avoid dehydration of the pH sensing membrane, which erodes the reaction time of the electrode. Electrodes are best stored with the tip immersed in the bridge electrolyte (KCl 3M). Most electrodes have a protective cap that can be filled with this solution before placing.

Specifications

CAS 7447-40-7
Quantity 250 mL
Odor Odorless
Molecular Formula ClK
MDL Number MFCD00011360
Physical Form Liquid
Concentration 224g KCl per 1L H2O
Color Colorless
pH >7
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Boiling Point 100°C
SMILES [Cl-].[K+]
For Use With (Equipment) pH Electrode
IUPAC Name potassium chloride
Solubility Soluble in water
Molecular Weight (g/mol) 74.55
PubChem CID 4873
ChEBI CHEBI:32588
Formula Weight 74.56g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium Chloride Electrolyte Standard Solution, 3M, Chem-Lab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.