Learn More
BINDER™ Electromechanical Door Lock with Pin
Product Code. 30238243 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB 65, KB 130, KB 260, KB 470
KB 720, KB 1060
KB PRO 130/260/470
KB PRO 260, KBF PRO 130/260/470
KB PRO 720/1060/1600
KBF 130, KBF 260, KBF 470
KBF 720, KBF 1060, KBF 1600
KBF PRO 720/1060/1600
Unit Size:
Each
Product Code. 30238243
Brand: BINDER™ 80122589
Only null left
Add to basket
Add to basket
Lockable door handle that can be opened with PIN code via controller
Door is locked with a PIN code via a controller to ensure secure access to samples.
Specifications
For Use With (Equipment) KBF 720, KBF 1060, KBF 1600
Material Steel
Type Locking Latch