Learn More
OHAUS™ Electronic Balance - NVT Series
Multi-Purpose Portable Balances Suitable for Everyday Weighing
Brand:  OHAUS™ 30456387
161.72 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 16600314
  • £241.00 / Each
Explore more special offers
Description
Description
  • Navigator offers essential weighing applications at a competitive price point.
  • Advanced weighing technology offers one-second stabilization for fast and accurate weighing results, improving operator efficiency, productivity and throughput.
  • Overload protection system that can withstand loads up to four times the rated capacity safeguards the balance.
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
OHAUS™ Electronic Balance - NVT Series > Model: NV1201