Learn More
OHAUS™ Electronic Balance - NVT Series
Multi-Purpose Portable Balances Suitable for Everyday Weighing
£166.00 - £422.00
Specifications
Connection Type | RS232, USB, or Ethernet (available as accessories) |
---|---|
Description | Menu and calibration lock switch, mechanical and software overload/underload protection, stability indicator, low battery indicator, auto shut-off, user-selectable print options, user-selectable communication settings |
Display Type | Backlit LCD |
For Use With (Application) | Weighing, Parts Counting, Percent Weighing, Checkweighing |
Electrical Requirements | AC adapter (included) or 4 C batteries (not included) |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
16315125
|
OHAUS™
30456391 |
NVT6201 |
£406.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16313488
|
OHAUS™
30456394 |
NVT12000 |
£265.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16474529
|
OHAUS™
30456388 |
NV2201 |
£254.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16620643
|
OHAUS™
30456382 |
NV222 |
£361.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16660304
|
OHAUS™
30456383 |
NV422 |
£381.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16670304
|
OHAUS™
30456384 |
NV622 |
£422.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16680304
|
OHAUS™
30456385 |
NV221 |
£184.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16690304
|
OHAUS™
30456386 |
NV621 |
£207.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16600314
|
OHAUS™
30456387 |
NV1201 |
£241.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16610314
|
OHAUS™
30456389 |
NVT2201 |
£288.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16620314
|
OHAUS™
30456390 |
NVT4201 |
£301.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16630314
|
OHAUS™
30456392 |
NVT2200 |
£207.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16640314
|
OHAUS™
30456393 |
NVT6200 |
£166.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Navigator offers essential weighing applications at a competitive price point.
- Advanced weighing technology offers one-second stabilization for fast and accurate weighing results, improving operator efficiency, productivity and throughput.
- Overload protection system that can withstand loads up to four times the rated capacity safeguards the balance.
Specifications
RS232, USB, or Ethernet (available as accessories) | |
Backlit LCD | |
AC adapter (included) or 4 C batteries (not included) |
Menu and calibration lock switch, mechanical and software overload/underload protection, stability indicator, low battery indicator, auto shut-off, user-selectable print options, user-selectable communication settings | |
Weighing, Parts Counting, Percent Weighing, Checkweighing |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
OHAUS™ Electronic Balance - NVT Series