Learn More
OHAUS™ Electronic Balance - PR Series
Designed for Routine Weighing Applications in Your Workplace
£710.00 - £1605.00
Specifications
Connection Type RS232 (one port included)
Description User-selectable environmental filters and brightness settings, auto-tare, auto-dim, user-selectable span calibration points, software lockout and reset menu, user-selectable communication settings and data print options, user-definable project and user IDs, software overload/underload indicator, stability indicator
Display Type Backlit LCD
Weighing Modes Milligram, Gram, Kilogram, Ounce, Pound, Carat, Pennyweight, Ounce Troy, Newton, & Grain
Length (English) 12.50 in.
View More Specs
Products 20
Product Code Brand Model Price Quantity & Availability  
Product Code Brand Model Price Quantity & Availability  
16357575
View Documents Promotion Details
OHAUS™
30430069
PR224
£1605.00
Each
16381678
View Documents Promotion Details
OHAUS™
30430076
PR4202/E
£965.00
Each
16389386
View Documents Promotion Details
OHAUS™
30492735
PR4202
£1110.00
Each
16506244
View Documents
OHAUS™
30430075
PR2202/E
£874.00
Each
16516244
View Documents Promotion Details
OHAUS™
30492734
PR2202
£979.00
Each
16680294
View Documents Promotion Details
OHAUS™
30430068
PR124/E
£1100.00
Each
16670294
View Documents Promotion Details
OHAUS™
30430067
PR124
£1225.00
Each
16690294
View Documents Promotion Details
OHAUS™
30430070
PR224/E
£1330.00
Each
16600304
View Documents Promotion Details
OHAUS™
30430071
PR223/E
£881.00
Each
16680314
View Documents
OHAUS™
30492730
PR223
£922.00
Each
16610304
View Documents Promotion Details
OHAUS™
30430072
PR423/E
£772.00
Each
16690314
View Documents
OHAUS™
30492731
PR423
£943.00
Each
16620304
View Documents Promotion Details
OHAUS™
30430073
PR523/E
£850.00
Each
16600324
View Documents Promotion Details
OHAUS™
30492732
PR523
£1085.00
Each
16630304
View Documents Promotion Details
OHAUS™
30430074
PR1602/E
£772.00
Each
16610324
View Documents Promotion Details
OHAUS™
30492733
PR1602
£948.00
Each
16640304
View Documents
OHAUS™
30430077
PR4201/E
£710.00
Each
16620324
View Documents
OHAUS™
30492736
PR4201
£886.00
Each
16650304
View Documents Promotion Details
OHAUS™
30430078
PR6201/E
£741.00
Each
16630324
View Documents Promotion Details
OHAUS™
30498176
PR6201
£906.00
Each
Description
Description
  • The PR is equipped with 3 essential weighing modes, RS-232 connectivity for data transfer and storage, and internal calibration (in select models), making it ideal for routine weighing applications.
  • Equipped with an easy-to-read, bright backlit display and a simple user interface, the PR is incredibly easy to operate, with almost no training required.
  • The PR's small footprint saves desktop space while providing a large weighing surface. The PR is durably constructed, and features a stainless steel pan to withstand day-to-day use in the workplace.
Specifications
Specifications
RS232 (one port included)
Backlit LCD
12.50 in.
7.90 in.
12.50 in.
AC Adapter (included)
User-selectable environmental filters and brightness settings, auto-tare, auto-dim, user-selectable span calibration points, software lockout and reset menu, user-selectable communication settings and data print options, user-definable project and user IDs, software overload/underload indicator, stability indicator
Milligram, Gram, Kilogram, Ounce, Pound, Carat, Pennyweight, Ounce Troy, Newton, & Grain
317.00 mm
201.00 mm
317.00 mm
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
OHAUS™ Electronic Balance - PR Series