missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ Solarus™ Electronic Burette
Digital, solar-powered display for easy reading of titration results. High accuracy with double seal PTFE compound piston.
Brand: Hirschmann™ 9392010
Description
- For sensitive manual titration
- Media conveying parts can be steam sterilized at 121°C
- Free rotation
- Media recirculating system for simple and rapid venting without reagent loss
- Solar panels being the power supply for the display
- Use within a temperature range of 4°C to 50°C
- Individual quality certificate with conformity certificate
Specifications
Autoclavable | |
10 mL | |
Manual Rotation and PC Connection | |
Electronic Buret |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction