Learn More
3M™ Electronic Test System Data Reader
The ETS Data Reader reads data from your main ETS unit and downloads to a PC.
Brand:  3M™ 7100194284
Additional Details : Weight : 0.14000kg
Product Code. 17781669
  • £401.00 / Each
Estimated Shipment: 02-09-2024
to see stock.
Add to basket
Description
Description
  • Service parts available to authorized entities only
  • Uses LED wireless connection to read data from ETS
  • Accessory to main ETS Sensing Unit
Specifications
Specifications
Reading Data From ETS Unit
ETS Sensing Unit (4308)
4309
Non-sterile