missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Electroporation Chambers: Petri Pulser Electrode
Petri Pulser Electrode is an immersion electrode for adherent cells in situ.
Brand: BTX™ 450130
This item is not returnable.
View return policy
Description
- Immersion electrode for adherent cells in situ
- Works in 35 mm Petri dishes
- Fits single well of 6-well plate
Specifications
Reusable electroporation applicator for 6-well plates or 35 mm petri dishe | |
Gemini X2, ECM 830, ECM 630, ECM 399, ECM 2001+ Generators | |
BTX Electroporation | |
300 V | |
Homogenous | |
0.5 mm | |
1 Ea. | |
2 mm | |
0.5 to 3 mL | |
0.17 kg | |
Banana Cables |
Mammalian Cell Transfection, Gene Therapy, Protein or Drug Delivery, Plant/Yeast | |
Gold-plated | |
Electroporation accessory | |
Petri Pulser | |
0.019 in. | |
1 μsec. to 35 msec. | |
0.078 in. | |
0.5 mm | |
0.37 lb. | |
Non-sterile | |
Exponential Decay Wave, Square Wave |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction