missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bibby Scientific™ Electrothermal™ Heating Element for EM/EME
Bibby Scientific Electrothermal Heating Element for EM/EME, Capacity: 250 mL, For Use With: 14314 EM Series Heating Mantles, Voltage: 230 V, Frequency: 50/60 Hz
£227.70 - £595.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
11738412
|
Bibby Scientific™
03012-55 |
1000 mL |
£440.00
Each |
Estimated Shipment: 17-04-2025
Log in to see stock. |
||||||
11748412
|
Bibby Scientific™
10086-37 |
2000 mL |
£595.00
Each |
Estimated Shipment: 17-04-2025
Log in to see stock. |
||||||
11768412
|
Bibby Scientific™
03012-85 |
5000 mL |
£429.00
Each |
Estimated Shipment: 17-04-2025
Log in to see stock. |
||||||
11876475
|
Bibby Scientific™
03012-35 |
250 mL |
£447.00
Each |
In Stock
Log in to see stock. |
||||||
11728412
|
Bibby Scientific™
03012-45 |
500 mL |
£227.70
Each |
In Stock
Log in to see stock. |
||||||
Description
Bibby Scientific Electrothermal Heating Element for EM/EME, Capacity: 250 mL, For Use With: 14314 EM Series Heating Mantles, Voltage: 230 V, Frequency: 50/60 Hz
Specifications
230 V | |
Heating Element | |
50/60 Hz |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bibby Scientific™ Electrothermal™ Heating Element for EM/EME