missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ELISA Systems™ Food Allergen Assay Accessories

Support the functionality of ELISA Systems™ Food Allergen Test Kits with ELISA Systems Food Allergen Assay Accessories.

Brand:  Thermo Scientific™ ESADDSOL

56.77 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 13275649

  • £53.50 / 25mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Support the functionality of ELISA Systems™ Food Allergen Test Kits with ELISA Systems Food Allergen Assay Accessories, including extraction and wash solutions. ELISA Systems Food Allergen Test Kits are a comprehensive range of enzyme-linked immunosorbent assays (ELISA), designed to detect and quantify the presence of specific proteins or residues in food products and environmental samples, to help you ensure compliance with food product labeling requirements.

ELISA Systems Food Allergen Test Kits come complete and ready-to-use. Solutions can be ordered separately, where required:
  • ELISA Systems Extraction Solution: Compatible with most of the ELISA Systems Food Allergen Test Kits, thereby reducing sample preparation time
  • ELISA Systems Extraction Solution Polyphenol: A special extraction solution for samples containing polyphenols, including dark chocolate, wine, fruit juices, herbs, and tannins. Already included with nut test kits
  • ELISA Systems Wash Solution


ELISA Systems Food Allergen Test Kits are optimized to deliver fast detection and quantification of specific food allergens:
  • Fast: Sample preparation to result in under 60 minutes for rapid test turnaround and actions
  • Simple: Ready-to-use liquid reagents and concentrates, plus common handling procedure for all kits, make ELISA Systems Assays easy to implement and run
  • Robust: Ready-to-use liquid controls and allergen-free non-control reagents mean reduced risk of cross contamination that could lead to false-positive results
  • Flexible: Use for qualitative or quantitative testing—a single kit for more applications
  • Sensitive: Detect proteins or protein residues, including heat stable proteins, at low levels for improved assurance of product safety
  • Supported: Our experienced technical team is ready to assist you and help mitigate delays in test reporting


Interpretation is based on the suggested extraction/dilution protocol. Results are for screening purposes. All results should be interpreted as part of a HACCP plan for food allergens. Any sample returning a positive result should be regarded as a presumptive result, and confirmation or further testing should be performed.

Not all products are available for sale in all territories. Please inquire.

ELISA Systems™ Food Allergen Test Kits are manufactured by ELISA Systems Pty Ltd.

ELISA Systems is a trademark of ELISA Systems Pty Ltd.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ELISA Systems Extraction Solution Polyphenol (20X concentrate)
ELISA (Standard)
Extraction Solution
25 mL Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers