missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Ellipso-Spoon and Spatula Samplers
Special curved shape prevents missed sample accumulation in corners.
£27.09 - £74.50
Product Code | Brand | Capacity (Metric) | Length (Metric) | Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Length (Metric) | Size | Price | Quantity & Availability | |||
12917320
|
Bel-Art™
H36809-0050 |
70 mL bowl | 50 cm | 28 x 65 mm bowl |
£74.50
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
15500132
|
Bel-Art™
H36807-0021 |
10 mL Bowl | 21 cm | 15 x 35 mm bowl |
£27.09
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
12907320
|
Bel-Art™
H36806-0015 |
10 mL Bowl | 15 cm | 15 x 35 mm bowl |
£29.75
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
15510132
|
Bel-Art™
H36808-0030 |
70 mL bowl | 30 cm | 28 x 65 mm bowl |
£57.12
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
12894801
|
Bel-Art™
H36809-0040 |
70 mL bowl | 40 cm | 28 x 65 mm bowl |
£72.25
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
Description
- Crystals, powders, granulated and solid substances, and thick liquids retrieved
- Includes spoon end and spatula end
- Made of polished 304 grade stainless steel
Specifications
Spoon and Spatula Sampler |