Learn More
Elma™ Elmasonic X-tra Flex 2 Ultrasonic Cleaner
Brand: Elma™ 1033342
This item is not returnable.
View return policy
Specifications
V2A Stainless Steel Casing, V4A Stainless Steel Tank | |
3 x 400 VAC/N/PE | |
1070 mm | |
3 x 400VAC/N/PE, 9300w | |
1500 mm | |
Elmasonic X-tra Line 1200 Flex 2 | |
9300 w | |
1450 mm |
Yes | |
112 L | |
1070 x 1450 x 1500 mm | |
Modular cleaning | |
40kg Capacity, 222 H x 436 W x 509mm D internal dimension Stainless Steel EKO basket, 8/8/1mm EKO basket mesh size, Stainless Steel cover, support frame | |
30°C to 80°C | |
400 kg |