Learn More
Elma™ Elmasonic X-tra Flex 2 Ultrasonic Cleaner
Brand: Elma™ 1033312
This item is not returnable.
View return policy
Specifications
V2A Stainless Steel Casing, V4A Stainless Steel Tank | |
1 x 230 to 240 VAC/N/PE | |
770 mm | |
1 x 230 to 240VAC/N/PE, 2300w | |
1100 mm | |
Elmasonic X-tra Line 300 Flex 2 | |
2300 w | |
500 mm |
Yes | |
32 L | |
770 x 500 x 1100 mm | |
Modular cleaning | |
20kg Capacity, 188 H x 248 W x 258mm D internal dimension Stainless Steel EKO basket, 8/8/1mm EKO basket mesh size, Stainless Steel cover, support frame | |
30°C to 80°C | |
80 kg |