Learn More
Elma™ Elmasonic X-tra Flex 2 Ultrasonic Cleaner
Brand: Elma™ 1033320
This item is not returnable.
View return policy
Specifications
V2A Stainless Steel Casing, V4A Stainless Steel Tank | |
3 x 400 VAC/N/PE | |
940 mm | |
3 x 400VAC/N/PE, 3900w | |
1200 mm | |
Elmasonic X-tra Line 550 Flex 2 | |
3900 w | |
1000 mm |
Yes | |
55 L | |
940 x 1000 x 1200 mm | |
Modular cleaning | |
20kg Capacity, 224 H x 248 W x 428mm D internal dimension Stainless Steel EKO basket, 8/8/1mm EKO basket mesh size, Stainless Steel cover, support frame | |
30°C to 80°C | |
180 kg |