Learn More
ELP3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522196
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: NT2D1, IMR32. Predicted reactivity: Mouse (96%), Zebrafish (92%), Drosophila (88%), Yeast (84%), Xenopus laevis (92%), Chicken (91%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Elongator complex protein 3 (ELP3) is a catalytic histone acetyltransferase subunit of the RNA polymerase II elongator complex, which is a component of the RNA polymerase II (Pol II) holoenzyme and is involved in transcriptional elongation. Elongator may play a role in chromatin remodeling and is involved in acetylation of histones H3 and probably H4. It may also have a methyltransferase activity.Specifications
ELP3 | |
Polyclonal | |
Unconjugated | |
ELP3 | |
2610507P14Rik; EC 2.3.1.48; elongation protein 3 homolog; elongator acetyltransferase complex subunit 3; elongator complex protein 3; ELP3; h KAT9; hELP3; KAT9; LOC100362548; Protein lysine acetyltransferase ELP3; rCG52086-like; tRNA uridine(34) acetyltransferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55140 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9H9T3 | |
ELP3 | |
Recombinant fragment corresponding to a region within amino acids 240 and 475 of ELP3 (Uniprot ID#Q9H9T3) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |