Learn More
Corning™ Elplasia™ Microcavity Plates
The only plates with a simple 'plug and play' protocol for scaffold-free, self-assembly spheroid formation at large volumes.
Brand:  Corning™ 4444
Additional Details : Weight : 4.17230kg
 View more versions of this product
Product Code. 16465299
  • £511.66 / Pack of 5
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
Description
Description
  • Novel well geometry and surface coatings, including Corning Ultra Low Attachment (ULA) featured on the round bottom plates
  • Corning ULA surface is hydrophilic, biologically inert and non-degradable, which promotes highly reproducible spheroid formation and easy harvesting
  • Corning Elplasia round bottom plates optimal for bulk spheroid formation, collection and expansion\
  • Round bottom plates feature Corning Ultra-Low Attachment surface and are available in 6-, 24- and 96-well formats
  • Corning Elplasia square bottom plates feature a surface with optical qualities suited for image analysis, making them an ideal solution for clonal selection and high magnification imaging of very small clusters
  • The Elplasia square plate is well suited for screening programs as it generates a high volume of spheroids per well, allowing for higher signals per well
  • Square bottom plates are plasma-treated for self-coating and come in 6-, 24-, 96- and 384-well formats
  • Spheroids may be formed and cultured for 21 or more days
  • Can produce between 79 – 15,000+ spheroids per well, depending on plate format, under one culture condition
  • The high volume of spheroids generated in each well increase signals per well without increasing spheroid size
  • Black opaque sidewalls to reduce well-to-well 'cross-talk'
  • Well suited for fluorescent/luminescent assays
  • Available in multiple formats, two well geometries and surface coating options
Specifications
Specifications
Clear
Polystyrene
Round
Polystyrene
Sterile
Microcavity Plate
Lid
Closure Included
Not Autoclavable
Black, Clear
Rectangular
Tissue Culture
6
5