missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tuttnauer ELV-D Vertical Autoclave
Shop All Tuttnauer Products
Click to view available options
Capacity (Metric):
31 L
65 L
85 L
160 L
Features:
Single Phase
Single Phase, Water Reservoir
Three Phase
Three Phase, BioHazard Filter
Three Phase, Water Reservoir
Water Reservoir
Unit Size:
Each
Description
Designed for your laboratory applications. The 5075ELV-D has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, Fo control, and more.
- Advanced Control System - Take Advantage of Tuttnauer's state-of-the-art control system with multi-color display
- Safe - Automatic pressure locking system. Door will not open when the chamber is overpressured
- Optional Internal Printing - Documents date, time, temperature and pressure
- Intuitive Multi-Color Display - Multi-color display for easier reading gives quick access to important information. Color is used to indicate the stage of the cycle
- High Quality Stainless Steel - 316L grade stainless steel chamber and door with electro-polish finish
- Diverse Load Types - The 5075ELV-D autoclave comes with 8 factory set cycle programs that cover the processing needs of most laboratories and research centers
Specifications
Specifications
| Capacity (English) | 14.3 gal. |
| Capacity (Metric) | 65 L |
| Depth (Metric) Chamber | 50 cm |
| Diameter (English) Chamber | 15.2 in. |
| Depth (English) Chamber | 20 in. |
| Diameter (Metric) Chamber | 38 cm |
| Length (English) | 32 in. |
| Length (Metric) | 80 cm |
| Model | 3850ELVWR-D |
| Product Type | Autoclave, Top-loading |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction