Learn More
Tuttnauer ELV-D Vertical Autoclave
The ELV-D Vertical Autoclave has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, biohazard and waste sterilization, Fo control, and more.
Brand: Tuttnauer 5075ELVCPVGP-230-D
Description
Designed for your laboratory applications. The 5075ELV-D has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, Fo control, and more.
- Advanced Control System - Take Advantage of Tuttnauer's state-of-the-art control system with multi-color display
- Safe - Automatic pressure locking system. Door will not open when the chamber is overpressured
- Optional Internal Printing - Documents date, time, temperature and pressure
- Intuitive Multi-Color Display - Multi-color display for easier reading gives quick access to important information. Color is used to indicate the stage of the cycle
- High Quality Stainless Steel - 316L grade stainless steel chamber and door with electro-polish finish
- Diverse Load Types - The 5075ELV-D autoclave comes with 8 factory set cycle programs that cover the processing needs of most laboratories and research centers
Specifications
35.2 gal. | |
75 cm | |
30 in. | |
30.8 in. | |
5075ELV-D | |
221°F to 280°F | |
Single Phase, Cooling, Vacuum, Steam Generator, Printer |
160 L | |
20 in. | |
50 cm | |
77 cm | |
Autoclave, Top-loading | |
105°C to 138°C | |
Non-sterile |