Learn More
Motic™ ELWD Condenser for AE2000
Product Code. 11792348
Click to view available options
For Use With (Equipment):
AE2000 Microscopes
Unit Size:
Each
Product Code. 11792348
Brand: Motic™ 1101000201621
Only null left
Add to basket
Add to basket
Extra-long working distance condenser suitable for phase contrast. Motic™ ELWD Condenser for AE2000, when used with phase accessories, facilitates visualization of transparent specimens.
  • Concentrates and focuses light passing through the specimen.
  • Extra-long working-distance condenser.
  • Suitable for phase contrast.
Specifications
Description NA: 0.30
Product Type ELWD Condenser
For Use With (Equipment) AE2000 Microscopes
Working Distance 72 mm